1,4-Dichlorophthalazine ≥98%

Supplier: BeanTown Chemical

223230-1G 223230-5G
BT223230-1GEA 48 USD
BT223230-1G BT223230-5G
1,4-Dichlorophthalazine ≥98%
1,4-Dichlorophthalazine

CAS: 4752-10-7; EC No: 225-275-6; MDL No: MFCD00006909 Powder; Molecular Formula: C8H4Cl2N2; MW: 199.04 Melting Point: 160-162° Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₈H₄Cl₂N₂
MW: 199.04 g/mol
Melting Pt: 160…162 °C
MDL Number: MFCD00006909
CAS Number: 4752-10-7
EINECS: 225-275-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR