L-Lysine acetate salt ≥98%

Supplier: BeanTown Chemical

Synonyms: L-LYSINE, MONOACETATE, (2S)-2,6-diaminohexanoic acid, L-Lysine acetate, L-Lysine Acetate Salt

223180-100G 223180-25G 223180-500G
BT223180-100GEA 189 USD
BT223180-100G BT223180-25G BT223180-500G
L-Lysine acetate salt ≥98%
L-Lysine acetate salt

CAS: 57282-49-2; EC No: 260-664-4; MDL No: MFCD00039069; RTECS: OL5644200 Solid; Linear Formula: NH2(CH2)4CH(NH2)COOH·CH3COOH; Molecular Formula: C6H14N2O2·C2H4O2; MW: 206.24


  • RTECS: OL5644200
  • Solid

Formula: C₈H₁₈N₂O₄
MW: 206.242 g/mol
MDL Number: MFCD00039069
CAS Number: 57282-49-2
EINECS: 260-664-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR