2,6-Dichloronicotinic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid
223125-1G
223125-5G
BT223125-1GEA
52
USD
BT223125-1G
BT223125-5G
2,6-Dichloronicotinic acid ≥97%
2,6-Dichloronicotinic acid
CAS: 38496-18-3; EC No: 000-000-0; MDL No: MFCD00075583
Solid; Molecular Formula: C6H3Cl2NO2; MW: 192.00
Melting Point: 140-143°
- Solid
Formula:
C₆H₃Cl₂NO₂ MW: 192 g/mol Melting Pt: 140…143 °C |
MDL Number:
MFCD00075583 CAS Number: 38496-18-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...