Benzyltriethylammonium chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Benzyl triethylammonium chloride
223100-100G
223100-500G
BT223100-100GEA
34
USD
BT223100-100G
BT223100-500G
Benzyltriethylammonium chloride ≥98%
Benzyltriethylammonium chloride
CAS: 56-37-1; EC No: 200-270-1; MDL No: MFCD00011824; RTECS: BO8275000
Crystalline; Molecular Formula: C13H22ClN; MW: 227.77
Melting Point: 190-192° (decomposes)
Hygroscopic
- RTECS: BO8275000
- Hygroscopic
- Crystalline
Formula:
C₁₃H₂₂ClN MW: 227.78 g/mol Melting Pt: 190…192 °C (dec.) |
MDL Number:
MFCD00011824 CAS Number: 56-37-1 EINECS: 200-270-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...