4-Ethynylpyridine hydrochloride ≥97%
Supplier: BeanTown Chemical
223075-1G
223075-5G
BT223075-1GEA
97
USD
BT223075-1G
BT223075-5G
4-Ethynylpyridine hydrochloride ≥97%
4-Ethynylpyridine hydrochloride
CAS: 352530-29-1; MDL No: MFCD03093912
Solid; Molecular Formula: C7H5N·HCl ; MW: 139.58
Melting Point: 150° (decomposes)
- Solid
Formula:
C₇H₆ClN MW: 139.58 g/mol Melting Pt: 150 °C (dec.) |
MDL Number:
MFCD03093912 CAS Number: 352530-29-1 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...