1,8-Dibromooctane ≥98%
Supplier: BeanTown Chemical
CAS: 4549-32-0; EC No: 224-912-5; MDL No: MFCD00000277
Liquid; Linear Formula: Br(CH2)8Br; Molecular Formula: C8H16Br2; MW: 272.02
Melting Point: 12-16°; Boiling Point: 270-272°; Flash point:
- Refractive index: 1.498
- Liquid
Formula:
BrH₂C(CH₂)₆CH₂Br Boiling Pt: 270…272 °C Melting Pt: 12…16 °C Density: 1.477 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00000277 CAS Number: 4549-32-0 EINECS: 224-912-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...