Methyl cellulose, 400 cP (2% H₂O)
Supplier: BeanTown Chemical
Synonyms:
Cellulose methyl ether
223060-100G
223060-2.5KG
223060-500G
BT223060-100GEA
46.65
USD
BT223060-100G
BT223060-2.5KG
BT223060-500G
Methyl cellulose, 400 cP (2% H₂O)
Methyl cellulose
CAS: 9004-67-5; MDL No: MFCD00081763
Powder
- Powder
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 Merck Index: 14,6040 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...