D-(+)-Mannose ≥99%
Supplier: BeanTown Chemical
Synonyms:
(2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
223055-100G
223055-25G
223055-500G
BT223055-100GEA
90
USD
BT223055-100G
BT223055-25G
BT223055-500G
D-(+)-Mannose ≥99%
D-(+)-Mannose
CAS: 3458-28-4; EC No: 222-392-4; MDL No: MFCD00064122
Solid; Molecular Formula: C6H12O6; MW: 180.16
Melting Point: 133-140°
Optical Rotation: [α]20/D +13.8±0.5°, c = 10% in water
Hygroscopic
- Optical rotation: [α]20/D +13.8±0.5°, c = 10% in water
- Hygroscopic
- Solid
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 376 Melting Pt: 133…140 °C |
MDL Number:
MFCD00064122 CAS Number: 3458-28-4 EINECS: 222-392-4 Merck Index: 13,05772 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...