Ethyl-5-bromothiophene-2-carboxylate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 5-bromothiophene-2-carboxylate
223015-1G
223015-25G
223015-5G
BT223015-1GEA
50
USD
BT223015-1G
BT223015-25G
BT223015-5G
Ethyl-5-bromothiophene-2-carboxylate ≥99%
Ethyl 5-bromothiophene-2-carboxylate
CAS: 5751-83-7; MDL No: MFCD02683089
Liquid; Molecular Formula: C7H7BrO2S; MW: 235.10
Boiling Point: 94-96°/4 mmHg
- Liquid
Formula:
C₇H₇BrO₂S MW: 235.10 g/mol Boiling Pt: 94…96 °C/4 mmHg |
MDL Number:
MFCD02683089 CAS Number: 5751-83-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...