Copper ≥99.95% (trace metals basis), foil not certified pinhole-free, Thickness 0.015 mm
Supplier: BeanTown Chemical
222980-25X25MM
222980-50X50MM
BT222980-25X25EA
134
USD
BT222980-25X25
BT222980-50X50
Copper ≥99.95% (trace metals basis), foil not certified pinhole-free, Thickness 0.015 mm
Copper
CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965
Solid; Molecular Formula: Cu; MW: 63.55
Melting Point: 1083.4°; Boiling Point: 2567°
Density (g/mL): 8.94
- Solid
Formula:
Cu Boiling Pt: 2567 °C Melting Pt: 1083.4 °C Density: 8.94 g/mL |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...