Pyridine-2,6-dicarboxylic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA
222955-100G
222955-25G
222955-500G
BT222955-100GEA
69
USD
BT222955-100G
BT222955-25G
BT222955-500G
Pyridine-2,6-dicarboxylic acid ≥98%
Pyridine-2,6-dicarboxylic acid
CAS: 499-83-2; EC No: 207-894-3; MDL No: MFCD00006299
Crystalline; Molecular Formula: C7H5NO4; MW: 167.12
Melting Point: 248-250° (decomposes); Flash point: 188°C (370°F)
- Crystalline
Formula:
C₇H₅NO₄ MW: 167.12 g/mol Melting Pt: 248…250 °C (dec.) Flash Pt: 188 °C (370 °F) |
MDL Number:
MFCD00006299 CAS Number: 499-83-2 EINECS: 207-894-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...