2-Amino-3,5-diiodopyridine ≥95%

Supplier: BeanTown Chemical

Synonyms: 3,5-Diiodo-2-pyridylamine, 3,5-Diiodo-2-Pyridinamine, (3,5-Diiodo-2-Pyridyl)Amine, 3,5-Diiodopyridin-2-amine, 2-amino-3,5-diiodopyridine

222935-1G 222935-250MG
BT222935-1GEA 187 USD
BT222935-1G BT222935-250MG
2-Amino-3,5-diiodopyridine ≥95%
2-Amino-3,5-diiodopyridine

CAS: 23597-15-1; MDL No: MFCD02663852 Solid; Molecular Formula: C5H4I2N2; MW: 345.91 Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₅H₄I₂N₂
MW: 345.91 g/mol
MDL Number: MFCD02663852
CAS Number: 23597-15-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR