Hematoxylin ≥96% (dry weight)
Supplier: BeanTown Chemical
222930-100G
222930-25G
222930-5G
BT222930-100GEA
235
USD
BT222930-100G
BT222930-25G
BT222930-5G
Hematoxylin ≥96% (dry weight)
Hematoxylin
CAS: 517-28-2; EC No: 208-237-3; MDL No: MFCD00078111; RTECS: MH7875000
Powder; Molecular Formula: C16H14O6·xH2O; MW: 302.29 (anhydrous)
Melting Point: 200° (decomposes)
Light Sensitive
- RTECS: MH7875000
- Light Sensitive
- Powder
Formula:
C₁₆H₁₄O₆ Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 EINECS: 208-237-3 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...