6-Aminobenzothiazole ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Amino-1,3-benzothiazole, Benzothiazol-6-ylamine

222925-1G 222925-25G 222925-5G
BT222925-1GEA 65 USD
BT222925-1G BT222925-25G BT222925-5G
6-Aminobenzothiazole ≥98%
6-Aminobenzothiazole

CAS: 533-30-2; EC No: 208-559-4; MDL No: MFCD00015461; RTECS: DL1050200 Solid; Molecular Formula: C7H6N2S; MW: 150.20 Melting Point: 87-91°


  • RTECS: DL1050200
  • Solid

Formula: C₇H₆N₂S
MW: 150.2 g/mol
Melting Pt: 87…91 °C
MDL Number: MFCD00015461
CAS Number: 533-30-2
EINECS: 208-559-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR