1-Octanol ≥99%

Supplier: BeanTown Chemical

Synonyms: n-Octanol

222920-10L 222920-2.5L 222920-500ML 222920-19L 222920-57L
BT222920-10LEA 293.42 USD
BT222920-10L BT222920-2.5L BT222920-500ML BT222920-19L BT222920-57L
1-Octanol ≥99%
1-Octanol

CAS: 111-87-5; EC No: 203-917-6; MDL No: MFCD00002988; RTECS: RH6550000 Liquid; Linear Formula: CH3(CH2)7OH; Molecular Formula: C8H18O; MW: 130.23 Melting Point: -15°; Boiling Point: 196°; Flash point: 80°C (176°F) Density (g/mL): 0.827; Refractive Index: 1.429


  • RTECS: RH6550000
  • Refractive index: 1.429
  • Liquid

Formula: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Boiling Pt: 196 °C
Melting Pt: –15 °C
Density: 0.827 g/mL
Flash Pt: 80 °C (176 °F)
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR