Sodium ethyl-2-sulfolaurate

Supplier: BeanTown Chemical

Synonyms: Dodecanoic acid, 2-sulphoethyl ester, sodium salt, Sodium lauroyl isethionate, Lauroyl isethionate, sodium salt

222910-1G 222910-5G
BT222910-1GEA 162 USD
BT222910-1G BT222910-5G
Sodium ethyl-2-sulfolaurate
Sodium ethyl-2-sulfolaurate

CAS: 7381-01-3; EC No: 230-949-8 Solid; Molecular Formula: C14H27NaO5S; MW: 330.42


  • Solid

Formula: C₁₄H₂₇NaO₅S
MW: 330.415 g/mol
MDL Number: MFCD00047789
CAS Number: 7381-01-3
EINECS: 230-949-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR