Sodium bisulfite ACS

Supplier: BeanTown Chemical

Synonyms: Sodium hydrogen sulphite

222875-100G 222875-2.5KG 222875-500G
BT222875-100GEA 31.15 USD
BT222875-100G BT222875-2.5KG BT222875-500G
Sodium bisulfite ACS
Sodium bisulphite

CAS: 7631-90-5 Powder; Linear Formula: NaHSO3; MW: 104.06 Melting Point: 150° Density (g/mL): 1.48 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Powder

Formula: HNaO₃S
MW: 104.06 g/mol
Melting Pt: 150 °C
Density: 1.48 g/mL
MDL Number: MFCD00167602
CAS Number: 7631-90-5
UN: 2693
ADR: 8,III

Order Now

Specification Test Results

Assay (SO2) 58-99%
Insoluble matter ≤0.005%
Chloride (Cl) ≤0.02%
Arsenic (As) ≤0.0001%
Heavy metals (as Pb) ≤0.001%
Iron (Fe) ≤0.002%
Appearance White granules

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR