Dipotassium hydrogen orthophosphate trihydrate

Supplier: BeanTown Chemical

Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate

222860-1KG 222860-250G
BT222860-1KGEA 172 USD
BT222860-1KG BT222860-250G
Dipotassium hydrogen orthophosphate trihydrate
Dipotassium hydrogen orthophosphate trihydrate

CAS: 16788-57-1; EC No: 231-834-5; MDL No: MFCD00149926 Crystalline/Powder; Linear Formula: K2HPO4·3H2O; MW: 228.23 Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR