Pyridinium chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pyridine hydrochloride, Pyridinehydrochloride, Pyridine HCl
222845-1KG
222845-250G
222845-50G
BT222845-1KGEA
254
USD
BT222845-1KG
BT222845-250G
BT222845-50G
Pyridinium chloride ≥98%
Pyridinium chloride
CAS: 628-13-7; EC No: 211-027-4; MDL No: MFCD00012802; RTECS: UT4375000
Powder; Molecular Formula: C5H5N·HCl; MW: 115.56
Melting Point: 145-147°; Boiling Point: 222-224°
Hygroscopic
- RTECS: UT4375000
- Hygroscopic
- Powder
Formula:
C₅H₆ClN Boiling Pt: 222…224 °C Melting Pt: 145…147 °C |
MDL Number:
MFCD00012802 CAS Number: 628-13-7 EINECS: 211-027-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...