Aluminium oxide, γ-α-phase ≥99.98%

Supplier: BeanTown Chemical

222815-100G 222815-25G 222815-500G
BT222815-100GEA 110 USD
BT222815-100G BT222815-25G BT222815-500G
Aluminium oxide, γ-α-phase ≥99.98%
Aluminium oxide, γ-α-phase

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 0.01-0.02 Micron Powder, S.A. 100 m2/g ; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • 0.01-0.02 Micron Powder, S.A. 100 m²/g

Formula: Al₂O₃
MW: 101.96 g/mol
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Assay 99.98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR