Tris(3-methylphenyl)phosphine ≥98%
Supplier: BeanTown Chemical
222745-1G
222745-25G
222745-5G
BT222745-1GEA
18
USD
BT222745-1G
BT222745-25G
BT222745-5G
Tris(3-methylphenyl)phosphine ≥98%
Tris(3-methylphenyl)phosphine
CAS: 6224-63-1; EC No: 228-312-4; MDL No: MFCD00008532
Powder; Molecular Formula: C21H21P; MW: 304.37
Melting Point: 99-101°
- Powder
Formula:
C₂₁H₂₁P Melting Pt: 99…101 °C |
MDL Number:
MFCD00008532 CAS Number: 6224-63-1 EINECS: 228-312-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...