1-Bromo-3-methoxypropane ≥98%
Supplier: BeanTown Chemical
CAS: 36865-41-5; MDL No: MFCD02258473
UN No: UN2344; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3OCH2CH2CH2Br; Molecular Formula: C4H9BrO; MW: 153.02
Boiling Point: 132°; Flash Point:
- Liquid
Formula:
C₄H₉BrO Boiling Pt: 132 °C Density: 1.37 g/mL Flash Pt: <60 °C (<140 °F) |
MDL Number:
MFCD02258473 CAS Number: 36865-41-5 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...