≥98%

Supplier: BeanTown Chemical

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

222700-1KG 222700-250G 222700-5KG
BT222700-1KGEA 115.04 USD
BT222700-1KG BT222700-250G BT222700-5KG
≥98%
di-Potassium hydrogen phosphate

CAS: 7758-11-4; EC No: 231-834-5; MDL No: MFCD00011383; RTECS: TC5580000 Granular; Linear Formula: K2HPO4; MW: 174.18 Melting Point: 340° Density (g/mL): 2.3 Hygroscopic


  • RTECS: TC5580000
  • Hygroscopic
  • Granular

Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Density: 2.3 g/mL
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR