2-Ethylcaproic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Ethylhexanoic acid
222635-100ML
222635-2.5L
222635-500ML
BT222635-100MLEA
24
USD
BT222635-100ML
BT222635-2.5L
BT222635-500ML
2-Ethylcaproic acid ≥99%
2-Ethylcaproic acid
CAS: 149-57-5; EC No: 205-743-6; MDL No: MFCD00002675; RTECS: MO7700000
Liquid; Molecular Formula: C8H16O2; MW: 144.21
Boiling Point: 228°; Flash point: 114°C (237°F)
Density (g/mL): 0.903; Refractive Index: 1.425
- RTECS: MO7700000
- Refractive index: 1.425
- Liquid
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Boiling Pt: 228 °C Density: 0.903 g/mL Flash Pt: 114 °C (237 °F) |
MDL Number:
MFCD00002675 CAS Number: 149-57-5 EINECS: 205-743-6 |
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...