1,2,3,4-Tetrahydroquinoline ≥98%
Supplier: BeanTown Chemical
222600-100G
222600-500G
BT222600-100GEA
40
USD
BT222600-100G
BT222600-500G
1,2,3,4-Tetrahydroquinoline ≥98%
1,2,3,4-Tetrahydroquinoline
CAS: 635-46-1; EC No: 211-237-6; MDL No: MFCD00006693
Liquid; Molecular Formula: C9H11N; MW: 133.19
Melting Point: 9-14°; Boiling Point: 113-117°/10 mmHg; Flash point: 101°C (214°F)
Density (g/mL): 1.061; Refractive Index: 1.593
- Refractive index: 1.593
- Liquid
Formula:
C₉H₁₁N MW: 133.19 g/mol Boiling Pt: 113…117 °C/10 mmHg Melting Pt: 9…14 °C Density: 1.061 g/mL Flash Pt: 101 °C (214 °F) |
MDL Number:
MFCD00006693 CAS Number: 635-46-1 EINECS: 211-237-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...