1,4-Butanediol diglycidyl ether ≥96%

Supplier: BeanTown Chemical

Synonyms: Araldite RD-2, 1,4-Bis(oxiran-2-ylmethoxy)butane

222590-10G 222590-50G
BT222590-10GEA 58 USD
BT222590-10G BT222590-50G
1,4-Butanediol diglycidyl ether ≥96%
1,4-Butanediol diglycidyl ether

CAS: 2425-79-8; EC No: 219-371-7; MDL No: MFCD00005146; RTECS: EJ5100000 Liquid; Molecular Formula: C10H18O4; MW: 202.25 Boiling Point: 266°; Flash point: 110°C (230°F) Density (g/mL): 1.1; Refractive Index: 1.453 Hygroscopic


  • RTECS: EJ5100000
  • Refractive index: 1.453
  • Hygroscopic
  • Liquid

Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 266 °C
Density: 1.1 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00005146
CAS Number: 2425-79-8
EINECS: 219-371-7

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR