N-Carbobenzyloxy-D-proline ≥98%
Supplier: BeanTown Chemical
222580-1G
222580-25G
222580-5G
BT222580-1GEA
26
USD
BT222580-1G
BT222580-25G
BT222580-5G
N-Carbobenzyloxy-D-proline ≥98%
N-Carbobenzyloxy-D-proline
CAS: 6404-31-5; EC No: 229-021-5; MDL No: MFCD00063228
Solid; Molecular Formula: C13H15NO4 ; MW: 249.26
Melting Point: 76-78°
Optical Rotation: [α]25/D +40.2°, c = 2 in ethanol
- Optical rotation: [α]25/D +40.2°, c = 2 in ethanol
- Solid
Formula:
C₁₃H₁₅NO₄ Melting Pt: 76…78 °C |
MDL Number:
MFCD00063228 CAS Number: 6404-31-5 EINECS: 229-021-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...