Sodium thiocyanate ≥98.0% ACS
Supplier: BeanTown Chemical
222545-100G
222545-2KG
222545-500G
BT222545-100GEA
48.25
USD
BT222545-100G
BT222545-2KG
BT222545-500G
Sodium thiocyanate ≥98.0% ACS
Sodium thiocyanate
CAS: 540-72-7; EC No: 208-754-4; MDL No: MFCD00011123; RTECS: XL2275000
Solid; Linear Formula: NaSCN; MW: 81.07
Melting Point: 287°
Density (g/mL): 1.74
Hygroscopic, Light Sensitive
- RTECS: XL2275000
- Hygroscopic, Light Sensitive
- Solid
Formula:
NaSCN Melting Pt: 287 °C Density: 1.74 g/mL |
MDL Number:
MFCD00011123 CAS Number: 540-72-7 EINECS: 208-754-4 |
Specification Test Results
Assay | 98.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...