Calcium chloride dihydrate 99,0-105,0 ACS

Supplier: BeanTown Chemical

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

222530-2KG 222530-500G
BT222530-2KGEA 144.85 USD
BT222530-2KG BT222530-500G
Calcium chloride dihydrate 99,0-105,0 ACS
Calcium chloride dihydrate

CAS: 10035-04-8; EC No: 233-140-8; MDL No: MFCD00149613 Powder; Molecular Formula: CaCl2·2H2O; MW: 147.01 Melting Point: 176° (decomposes) Hygroscopic


  • Hygroscopic
  • Powder

Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176 (dec.)
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay 99.0-105.0%
pH of 5% solution at 25°C 4.5-9.2
Heavy metals (as Pb) ≤5ppm
Insoluble matter ≤0.01%
Oxidizing substances ≤0.003%
Sulfate (SO4) ≤0.01%
Ammonium (NH4) ≤0.005%
Barium (Ba) ≤0.005%
Iron (Fe) ≤0.001%
Magnesium (Mg) ≤0.005%
Potassium (K) ≤0.01%
Sodium (Na) ≤0.02%
Strontium (Sr) ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR