L-(-)-tert-Leucine (H-Tle-OH) ≥99%
Supplier: BeanTown Chemical
Synonyms:
(S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH
222505-1G
222505-25G
222505-5G
BT222505-1GEA
46
USD
BT222505-1G
BT222505-25G
BT222505-5G
L-(-)-tert-Leucine (H-Tle-OH) ≥99%
L(-)-tert-Leucine (H-Tle-OH)
CAS: 20859-02-3; EC No: 424-750-7; MDL No: MFCD00064218
Powder; Molecular Formula: C6H13NO2; MW: 131.17
Melting Point:
- Optical rotation: [α]20/D -9.5°, c = 3 in water
- Powder
Formula:
C₆H₁₃NO₂ MW: 131.2 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00064218 CAS Number: 20859-02-3 EINECS: 424-750-7 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...