Ethyl-2-aminothiazole-4-carboxylate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 2-aminothiazole-4-carboxylate
222490-1G
222490-5G
BT222490-1GEA
37
USD
BT222490-1G
BT222490-5G
Ethyl-2-aminothiazole-4-carboxylate ≥98%
Ethyl-2-aminothiazole-4-carboxylate
CAS: 5398-36-7; MDL No: MFCD00619079
Solid; Molecular Formula: C6H8N2O2S; MW: 172.20
Melting Point: 177-181°
- Solid
Formula:
C₆H₈N₂O₂S MW: 172.21 g/mol Melting Pt: 177…181 °C |
MDL Number:
MFCD00619079 CAS Number: 5398-36-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...