N,N'-Bis(acryloyl)cystamine ≥98%

Supplier: BeanTown Chemical

222450-1G 222450-5G
BT222450-1GEA 70 USD
BT222450-1G BT222450-5G
N,N'-Bis(acryloyl)cystamine ≥98%
N,N'-Bis(acryloyl)cystamine

CAS: 60984-57-8; EC No: 262-546-8; MDL No: MFCD00036225 Powder; Molecular Formula: C10H16N2O2S2; MW: 260.37 Melting Point: 123-124° Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₁₀H₁₆N₂O₂S₂
Melting Pt: 123…124 °C
MDL Number: MFCD00036225
CAS Number: 60984-57-8
EINECS: 262-546-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR