2-(Ethylsulfonyl)imidazo[1,2-a]pyridine-3-sulfonamide
Supplier: BeanTown Chemical
222445-1G
222445-5G
BT222445-1GEA
49
USD
BT222445-1G
BT222445-5G
2-(Ethylsulfonyl)imidazo[1,2-a]pyridine-3-sulfonamide
2-(Ethylsulfonyl)imidazo[1,2-a]pyridine-3-sulfonamide
CAS: 141776-47-8
Solid; Molecular Formula: C9H11N3O4S2; MW: 289.33
- Solid
Formula:
C₉H₁₁N₃O₄S₂ |
CAS Number:
141776-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...