2,4,6-Triiodophenol ≥98%

Supplier: BeanTown Chemical

222440-100G 222440-25G 222440-5G
BT222440-100GEA 309 USD
BT222440-100G BT222440-25G BT222440-5G
2,4,6-Triiodophenol ≥98%
2,4,6-Triiodophenol

CAS: 609-23-4; EC No: 210-186-7; MDL No: MFCD00002179; RTECS: SN2800000 Solid; Molecular Formula: C6H3I3O; MW: 471.80 Melting Point: 157-159° Light Sensitive


  • RTECS: SN2800000
  • Light Sensitive
  • Solid

Formula: C₆H₃I₃O
MW: 471.8 g/mol
Melting Pt: 157…159 °C
MDL Number: MFCD00002179
CAS Number: 609-23-4
EINECS: 210-186-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR