Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 4,000

Supplier: BeanTown Chemical

222365-1L 222365-250ML 222365-50ML
BT222365-1LEA 121 USD
BT222365-1L BT222365-250ML BT222365-50ML
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. ∼ 4,000
Polydimethylsiloxane, trimethylsiloxy terminated

CAS: 9016-00-6; MDL No: MFCD00148360; RTECS: TQ2690000 Liquid; Linear Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 ; MW ≈4,000 Melting Point: -40°; Boiling Point: 225°; Flash point: 285°C (545°F) Density (g/mL): 0.96; Refractive Index: 1.401


  • Refractive Index: 1.401

Formula: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Boiling Pt: >225 °C
Melting Pt: –40 °C
Density: 0.96 g/ml
Flash Pt: 285 °C
MDL Number: MFCD00148360
CAS Number: 9016-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR