2-Ethyl-4-methylimidazole ≥96%

Supplier: BeanTown Chemical

Synonyms: 2-Ethyl-4-methyl-1H-imidazole

222355-100G 222355-25G
BT222355-100GEA 83 USD
BT222355-100G BT222355-25G
2-Ethyl-4-methylimidazole ≥96%
2-Ethyl-4-methylimidazole

CAS: 931-36-2; EC No: 213-234-5; MDL No: MFCD00005193; RTECS: NI6147500 Solid; Molecular Formula: C6H10N2; MW: 110.16 Melting Point: 47-54°; Boiling Point: 292-295°; Flash point: 137°C (279°F) Density (g/mL): 0.975; Refractive Index: 1.5


  • RTECS: NI6147500
  • Refractive index: 1.5
  • Solid

Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 292…295 °C
Melting Pt: 47…54 °C
Density: 0.975 g/mL
Flash Pt: 137 °C (279 °F)
MDL Number: MFCD00005193
CAS Number: 931-36-2
EINECS: 213-234-5

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR