Tripropylene glycol (mixture of isomers)
Supplier: BeanTown Chemical
Synonyms:
Tripropylene glycol
222315-1L
222315-250ML
222315-4L
BT222315-1LEA
43
USD
BT222315-1L
BT222315-250ML
BT222315-4L
Tripropylene glycol (mixture of isomers)
Tripropylene glycol (mixture of isomers)
CAS: 24800-44-0; EC No: 246-466-0; MDL No: MFCD00014405
Liquid; Molecular Formula: C9H20O4; MW: 192.25
Melting Point:
- Refractive index: 1.444
- Liquid
Formula:
C₉H₂₀O₄ MW: 192.26 g/mol Boiling Pt: 273 °C Melting Pt: <–20 °C Density: 1.021 g/mL Flash Pt: 145 °C (293 °F) |
MDL Number:
MFCD00014405 CAS Number: 24800-44-0 EINECS: 246-466-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...