Platinum rhodium alloy (90:10 w%) ≥99.9% (trace metals basis), foil, Thickness 0.1 mm

Supplier: BeanTown Chemical

222310-25X25MM 222310-50X50MM
BT222310-25X25EA 359 USD
BT222310-25X25 BT222310-50X50
Platinum rhodium alloy (90:10 w%) ≥99.9% (trace metals basis), foil, Thickness 0.1 mm
Platinum rhodium alloy (90:10 w%)

MDL No: MFCD00198186 Solid; Formula: Pt:Rh; 90:10 wt%


  • Solid

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR