Triethyl phosphate ≥99%

Supplier: BeanTown Chemical

222290-100G 222290-25G 222290-500G
BT222290-100GEA 66.92 USD
BT222290-100G BT222290-25G BT222290-500G
Triethyl phosphate ≥99%
Triethyl phosphate

CAS: 78-40-0; EC No: 201-114-5; MDL No: MFCD00009077 Liquid; Linear Formula: (CH3CH2O)3P(O); Molecular Formula: C6H15O4P; MW: 182.16 Melting Point: -56°; Boiling Point: 215°; Flash point: 115°C (240°F) Density (g/mL): 1.072 ; Refractive Index: 1.403


  • Refractive index: 1.403
  • Liquid

Formula: C₆H₁₅O₄P
MW: 182.16 g/mol
Boiling Pt: 215 °C
Melting Pt: –56 °C
Density: 1.072 g/mL
Flash Pt: 115 °C (240 °F)
MDL Number: MFCD00009077
CAS Number: 78-40-0
EINECS: 201-114-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR