Magnesium fluoride ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

222270-1G 222270-5G
BT222270-1GEA 30 USD
BT222270-1G BT222270-5G
Magnesium fluoride ≥99.99% (trace metals basis)
Magnesium fluoride

CAS: 7783-40-6; EC No: 231-995-1; MDL No: MFCD00011108; RTECS: OM3325000 Powder; Linear Formula: MgF2; MW: 62.30 Melting Point: 1261° Density (g/mL): 3.148


  • RTECS: OM3325000
  • Powder

Formula: MgF₂
MW: 62.3 g/mol
Melting Pt: 1261 °C
Density: 3.148 g/mL
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR