Perfluorotri-n-butylamine, Mass Spec Standard

Supplier: BeanTown Chemical

222260-1G 222260-25G 222260-5G
BT222260-1GEA 61.12 USD
BT222260-1G BT222260-25G BT222260-5G
Perfluorotri-n-butylamine, Mass Spec Standard
Standards Organic Standards

EC No: 206-223-1
RTECS: YA1000000
GHS symbol: GHS07
Refractive Index: 1.289
Beilstein: 1813883

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR