(S)-(+)-β-Aminophenethyl alcohol ≥98%
Supplier: BeanTown Chemical
Synonyms:
(S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol
222255-1G
222255-25G
222255-5G
BT222255-1GEA
43
USD
BT222255-1G
BT222255-25G
BT222255-5G
(S)-(+)-β-Aminophenethyl alcohol ≥98%
(S)-(+)-β-Aminophenethyl alcohol
CAS: 20989-17-7; MDL No: MFCD00064404
Crystalline/Powder; Molecular Formula: C8H11NO; MW: 137.18
Melting Point: 75-78°
Optical Rotation: [α]19/D +33°, c = 0.75 in 1M HCl
Air Sensitive
- Optical rotation: [α]19/D +33°, c = 0.75 in 1M HCl
- Air Sensitive
- Crystalline/Powder
Formula:
C₈H₁₁NO MW: 137.18 g/mol Melting Pt: 75…78 °C |
MDL Number:
MFCD00064404 CAS Number: 20989-17-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...