2-Mercapto-5-methoxybenzimidazole ≥99%

Supplier: BeanTown Chemical

222250-100G 222250-25G 222250-5G
BT222250-100GEA 384 USD
BT222250-100G BT222250-25G BT222250-5G
2-Mercapto-5-methoxybenzimidazole ≥99%
2-Mercapto-5-methoxybenzimidazole

CAS: 37052-78-1; EC No: 253-326-2; MDL No: MFCD00134581 Powder; Molecular Formula: C8H8N2OS; MW: 180.23 Melting Point: 261-263° Air Sensitive


  • Air Sensitive
  • Powder

Formula: C₈H₈N₂OS
Melting Pt: 261…263 °C
MDL Number: MFCD00134581
CAS Number: 37052-78-1
EINECS: 253-326-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR