Europium(III) fluoride, anhydrous ≥99.5%
Supplier: BeanTown Chemical
222230-1G
222230-25G
222230-5G
BT222230-1GEA
43
USD
BT222230-1G
BT222230-25G
BT222230-5G
Europium(III) fluoride, anhydrous ≥99.5%
Europium(III) fluoride
CAS: 13765-25-8; EC No: 237-368-9; MDL No: MFCD00010998
-60 Mesh Powder; Molecular Formula: EuF3; MW: 208.96
Boiling Point: 2280°
Hygroscopic
- Hygroscopic
- -60 Mesh Powder
Formula:
EuF₃ MW: 208.96 g/mol Boiling Pt: 2280 |
MDL Number:
MFCD00010998 CAS Number: 13765-25-8 EINECS: 237-368-9 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...