Triammonium citrate ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

222205-100G 222205-500G
BT222205-100GEA 39 USD
BT222205-100G BT222205-500G
Triammonium citrate ≥97%
Triammonium citrate

CAS: 3458-72-8; EC No: 222-394-5; MDL No: MFCD00036406 Crystalline/Powder; Molecular Formula: C6H17N3O7; MW: 243.22 Melting Point: 185° (decomposes)


  • Crystalline/Powder

Formula: C₆H₁₇N₃O₇
Melting Pt: 185 °C (dec.)
MDL Number: MFCD00036406
CAS Number: 3458-72-8
EINECS: 222-394-5

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR