Trityl alcohol ≥98%

Supplier: BeanTown Chemical

Synonyms: Triphenylmethanol, Triphenylcarbinol

222155-1KG 222155-250G 222155-50G
BT222155-1KGEA 464 USD
BT222155-1KG BT222155-250G BT222155-50G
Trityl alcohol ≥98%
Trityl alcohol

CAS: 76-84-6; EC No: 200-988-5; MDL No: MFCD00004445 Solid; Linear Formula: (C6H5)3COH; Molecular Formula: C19H16O; MW: 260.33 Melting Point: 160-163°; Boiling Point: 360°


  • Solid

Formula: C₁₉H₁₆O
MW: 260.34 g/mol
Boiling Pt: 360 °C
Melting Pt: 160…163 °C
MDL Number: MFCD00004445
CAS Number: 76-84-6
EINECS: 200-988-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR