Ethyl-2-chloronicotinate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

222140-10G 222140-250G 222140-50G
BT222140-10GEA 76 USD
BT222140-10G BT222140-250G BT222140-50G
Ethyl-2-chloronicotinate ≥98%
Ethyl-2-chloronicotinate

CAS: 1452-94-4; MDL No: MFCD00674129 Liquid; Molecular Formula: C8H8ClNO2; MW: 185.61


  • Liquid

Formula: C₈H₈ClNO₂
MDL Number: MFCD00674129
CAS Number: 1452-94-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR