Diethylamine hydrochloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Diethylammonium chloride
222135-1KG
222135-250G
222135-5KG
BT222135-1KGEA
82.06
USD
BT222135-1KG
BT222135-250G
BT222135-5KG
Diethylamine hydrochloride ≥99%
Diethylamine hydrochloride
CAS: 660-68-4; EC No: 211-541-9; MDL No: MFCD00012499; RTECS: IA3084000
Crystalline; Linear Formula: (CH3CH2)2NH·HCl; Molecular Formula: C4H11N·HCl; MW: 109.60
Melting Point: 227-230°
Hygroscopic
- RTECS: IA3084000
- Hygroscopic
- Crystalline
Formula:
C₄H₁₂ClN MW: 109.6 g/mol Melting Pt: 227…230 °C |
MDL Number:
MFCD00012499 CAS Number: 660-68-4 EINECS: 211-541-9 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...