Tris(2-ethylhexyl)phosphate

Supplier: BeanTown Chemical

Synonyms: Trioctyl phosphate, Phosphoric acid tris(2-ethylhexyl ester), Tris(2-ethylhexyl) phosphate

222125-1KG 222125-250G 222125-50G
BT222125-1KGEA 145.05 USD
BT222125-1KG BT222125-250G BT222125-50G
Tris(2-ethylhexyl)phosphate
Tris(2-ethylhexyl)phosphate

CAS: 78-42-2; EC No: 201-116-6; MDL No: MFCD00009491 Liquid; Linear Formula: [CH3(CH2)3CH(CH2CH3)CH2O]3P(O) ; Molecular Formula: C24H51O4P; MW: 434.64 Boiling Point: 216°/4 mmHg; Flash point: 207°C (405°F) Density (g/mL): 0.92; Refractive Index: 1.441


  • Refractive index: 1.441
  • Liquid

Formula: C₂₄H₅₁O₄P
MW: 434.64 g/mol
Boiling Pt: 216 °C/4 mmHg
Density: 0.92 g/mL
Flash Pt: 207 °C (405 °F)
MDL Number: MFCD00009491
CAS Number: 78-42-2
EINECS: 201-116-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR