Sodium cholate hydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: 3α,7α,12α-Trihydroxycholanic acid sodium salt hydrate, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate hydrate, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt hydrate, Cholic acid sodium salt hydrate, Sodium 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoate hydrate, 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt hydrate, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt hydrate

222120-10G 222120-50G
BT222120-10GEA 29 USD
BT222120-10G BT222120-50G
Sodium cholate hydrate ≥99%
Sodium cholate hydrate

CAS: 206986-87-0; MDL No: MFCD00064138; RTECS: FZ9600000 Solid; Molecular Formula: C24H39NaO5·xH2O; MW: 430.55 (anhydrous) Optical Rotation: [α]20/D +36±2°, c = 0.6% in ethanol (dry matter) Hygroscopic


  • RTECS: FZ9600000
  • Optical rotation: [α]20/D +36±2°, c = 0.6% in ethanol (dry matter)
  • Hygroscopic
  • Solid

Formula: C₂₄H₃₉NaO₅·H₂O
MW: 448.58 g/mol
MDL Number: MFCD00064138
CAS Number: 206986-87-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR